Tisk…

Anti-Cyclin M2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Dodavatel: Bioss
BS-6912R-A680
BOSSBS-6912R-A680EA 25430 CZK
BOSSBS-6912R-A680
Anti-Cyclin M2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Protilátky
Divalent metal cation transporter. Mediates transport of divalent metal cations in an order of Mg(2+) > Co(2+) > Mn(2+) > Sr(2+) > Ba(2+) > Cu(2+) > Fe(2+) (By similarity).

Type: Primary
Antigen: Cyclin M2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 51-150/875
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR