Tisk…
Anti-MAGEL2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Dodavatel: Bioss
BS-6828R-A647
BOSSBS-6828R-A647EA 25430 CZK
BOSSBS-6828R-A647
Anti-MAGEL2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Protilátky
Melanoma-associated antigen (MAGE) are completely silent in normal tissues, with the exception of male germ cells, and, for some of them, placenta. These antigens ought to be strictly tumor specific, expressed in tumor cells of various histological types. Because of their specific expression on tumor cells, these antigens are of particular interest for antitumor immunotherapy. Genes of the MAGE family direct the expression of tumor antigens that are recognized on a human melanoma by autologous cytolytic T lymphocytes. Though the function of MAGE is unknown, may play a role in embryonalMore Product Information