Tisk…

Anti-CTAG1B Rabbit Polyclonal Antibody (Cy5.5®)

Dodavatel: Bioss
BS-6806R-CY5.5
BOSSBS-6806R-CY5.5EA 25630 CZK
BOSSBS-6806R-CY5.5
Anti-CTAG1B Rabbit Polyclonal Antibody (Cy5.5®)
Protilátky
Expressed in testis and ovary and in a wide variety of cancers. Detected in uterine myometrium. Expressed from 18 weeks until birth in human fetal testis. In the adult testis, is strongly expressed in spermatogonia and in primary spermatocytes, but not in post-meiotic cells or in testicular somatic cells (at protein level).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CTAG1B
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR