Tisk…

Anti-SDHB Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Dodavatel: Bioss
BS-6650R-A555
BOSSBS-6650R-A555EA 25430 CZK
BOSSBS-6650R-A555
Anti-SDHB Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Protilátky
Iron-sulfur protein (IP) subunit of succinate dehydrogenase (SDH) that is involved in complex II of the mitochondrial electron transport chain and is responsible for transferring electrons from succinate to ubiquinone (coenzyme Q).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SDHB
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR