Tisk…
Anti-ADAMTS10 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Dodavatel: Bioss
BS-6588R-A680
BOSSBS-6588R-A680EA 25430 CZK
BOSSBS-6588R-A680
Anti-ADAMTS10 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Protilátky
ADAMTS10 is a member of the ADAMs family of proteinases with Thrombospondin motifs. The catalytic site of ADAMTS10 is typical of the metalloproteinase catalytic domains, with an HExxHxxxxxH sequence, perhaps giving these enzymes some shared specificity. ADAMTS10 is closest in homology to ADAMTS6, sharing 53% overall identity. Functional mutations in ADAMTS10 have been linked to Weill Marchesani syndrome, a connective tissue disorder marked by fibrillin 1 misprocessing. ADAMTS10 has also been reported to be over expressed in breast cancer tissues and cell lines.
Type: Primary
Antigen:More Product Information