Tisk…

Anti-DUT Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Dodavatel: Bioss
BS-6584R-A555
BOSSBS-6584R-A555EA 25430 CZK
BOSSBS-6584R-A555
Anti-DUT Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Protilátky
This enzyme is involved in nucleotide metabolism: it produces dUMP, the immediate precursor of thymidine nucleotides and it decreases the intracellular concentration of dUTP so that uracil cannot be incorporated into DNA.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DUT
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR