Tisk…

Anti-PPAT Rabbit Polyclonal Antibody (Cy3®)

Dodavatel: Bioss
BS-6359R-CY3
BOSSBS-6359R-CY3EA 25630 CZK
BOSSBS-6359R-CY3
Anti-PPAT Rabbit Polyclonal Antibody (Cy3®)
Protilátky
Phosphoribosyl pyrophosphate amidotransferase (PPAT) is a regulatory allosteric enzyme that catalyzes the first step of de novo purine nucleotide biosynthesis. The genes for PPAT and PAICS/AIRC, (a bifunctional enzyme catalyzing steps six and seven in the purine nucleotide biosynthesis pathway), are located in close proximity on chromosome 4.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PPAT
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR