Tisk…
Anti-ADSL Rabbit Polyclonal Antibody (Cy5®)
Dodavatel: Bioss
BS-6352R-CY5
BOSSBS-6352R-CY5EA 25630 CZK
BOSSBS-6352R-CY5
Anti-ADSL Rabbit Polyclonal Antibody (Cy5®)
Protilátky
Adenylsuccinate lyase is involved in both de novo synthesis of purines and formation of adenosine monophosphate from inosine monophosphate. It catalyzes two reactions in AMP biosynthesis: the removal of a fumarate from succinylaminoimidazole carboxamide (SAICA) ribotide to give aminoimidazole carboxamide ribotide (AICA) and removal of fumarate from adenylosuccinate to give AMP. Adenylosuccinase deficiency results in succinylpurinemic autism, psychomotor retardation, and , in some cases, growth retardation associated with muscle wasting and epilepsy. Two transcript variants encoding differentMore Product Information
Objednat
Produktová specifikace
Symbol antigenu ADSL
Název antigenu Adenylosuccinate Lyase
Environmentally Preferable
Synonyma antigenu PUR8_HUMAN.|Adenylosuccinase|ASL|Adenylosuccinate lyase|AMPS|ASase|ADSL
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Cy5®
Reaktivita Human,Rat,Mouse
Hostitel Rabbit
Izotyp IgG
Imunochemie Yes
Formulář liquid
Čištění Purified by Protein A
Koncentrace 1 μg/μl
Křížová adsorbce No
Skladovací pufr Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Skladovací teplota Store at 4°C for 12 months
Transportní teplota 4°C