Tisk…

Anti-IMPAD1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Dodavatel: Bioss
BS-6248R-A555
BOSSBS-6248R-A555EA 25430 CZK
BOSSBS-6248R-A555
Anti-IMPAD1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Protilátky
This gene encodes a member of the inositol monophosphatase family. The encoded protein is localized to the Golgi apparatus and catalyzes the hydrolysis of phosphoadenosine phosphate (PAP) to adenosine monophosphate (AMP). Mutations in this gene are a cause of GRAPP type chondrodysplasia with joint dislocations, and a pseudogene of this gene is located on the long arm of chromosome 1.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IMPAD1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR