Tisk…

Anti-Wnt8b Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Dodavatel: Bioss
BS-6245R-A750
BOSSBS-6245R-A750EA 25430 CZK
BOSSBS-6245R-A750
Anti-Wnt8b Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Protilátky
Ligand for members of the frizzled family of seven transmembrane receptors. May play an important role in the development and differentiation of certain forebrain structures, notably the hippocampus.

Type: Primary
Antigen: Wnt8b
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 131-230/351
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR