Tisk…

Anti-WNT8B Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Dodavatel: Bioss
BS-6245R-A488
BOSSBS-6245R-A488EA 25430 CZK
BOSSBS-6245R-A488
Anti-WNT8B Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Protilátky
Ligand for members of the frizzled family of seven transmembrane receptors. May play an important role in the development and differentiation of certain forebrain structures, notably the hippocampus.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: WNT8B
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR