Tisk…

Anti-REN Rabbit Polyclonal Antibody (Cy3®)

Dodavatel: Bioss
BS-6184R-CY3
BOSSBS-6184R-CY3EA 26260 CZK
BOSSBS-6184R-CY3
Anti-REN Rabbit Polyclonal Antibody (Cy3®)
Protilátky
Renin is a highly specific endopeptidase, whose only known function is to generate angiotensin I from angiotensinogen in the plasma, initiating a cascade of reactions that produce an elevation of blood pressure and increased sodium retention by the kidney.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: REN
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR