Tisk…

Anti-WNT4 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Dodavatel: Bioss
BS-6134R-FITC
BOSSBS-6134R-FITCEA 26260 CZK
BOSSBS-6134R-FITC
Anti-WNT4 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Protilátky
Ligand for members of the frizzled family of seven transmembrane receptors. Probable developmental protein. May be a signaling molecule which affects the development of discrete regions of tissues. Is likely to signal over only few cell diameters (By similarity). Overexpression may be associated with abnormal proliferation in human breast tissue.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: WNT4
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Produktová specifikace

Symbol antigenu WNT4
Název antigenu Wingless-type MMTV integration site family, member 4
Environmentally Preferable
Synonyma antigenu WNT-4|SERKAL
Druh protilátky Primární
Klonovalita Polyclonal
Spojení FITC (Fluorescein Isothiocyanate)
Reaktivita Human,Rat,Mouse
Hostitel Rabbit
Identifikační číslo genu 54361
Izotyp IgG
Imunochemie Yes
Imunogen 240-290/351
Formulář liquid
Čištění Purified by Protein A
Koncentrace 1 μg/μl
Křížová adsorbce No
Skladovací pufr Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Skladovací teplota Store at 4°C for 12 months
Transportní teplota 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR