Tisk…

Anti-WNT2 Rabbit Polyclonal Antibody (Cy3®)

Dodavatel: Bioss
BS-6133R-CY3
BOSSBS-6133R-CY3EA 26060 CZK
BOSSBS-6133R-CY3
Anti-WNT2 Rabbit Polyclonal Antibody (Cy3®)
Protilátky
Ligand for members of the frizzled family of seven transmembrane receptors. Probable developmental protein. May be a signaling molecule which affects the development of discrete regions of tissues. Is likely to signal over only few cell diameters.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: WNT2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Dog
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR