Tisk…

Anti-WNT8A Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Dodavatel: Bioss
BS-6129R-A647
BOSSBS-6129R-A647EA 25430 CZK
BOSSBS-6129R-A647
Anti-WNT8A Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Protilátky
Ligand for members of the frizzled family of seven transmembrane receptors. May play an important role in the development and differentiation of certain forebrain structures, notably the hippocampus.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: WNT8A
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Produktová specifikace

Symbol antigenu WNT8A
Environmentally Preferable
Název antigenu Wingless-type MMTV Integration Site Family, Member 8A
Synonyma antigenu Stra11|Wnt-8A|Wnt-8D|Wnt8d
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Alexa Fluor® 647
Reaktivita Human,Rat,Mouse
Hostitel Rabbit
Identifikační číslo genu 7478
Izotyp IgG
Imunochemie Yes
Imunogen 200-250/351
Formulář liquid
Čištění Purified by Protein A
Koncentrace 1 μg/μl
Křížová adsorbce No
Skladovací pufr Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Skladovací teplota Store at 4°C for 12 months
Transportní teplota 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR