Tisk…
Anti-POU6F2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Dodavatel: Bioss
BS-6084R-A488
BOSSBS-6084R-A488EA 25430 CZK
BOSSBS-6084R-A488
Anti-POU6F2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Protilátky
This gene encodes a member of the POU protein family characterized by the presence of a bipartite DNA binding domain, consisting of a POU-specific domain and a homeodomain, separated by a variable polylinker. The DNA binding domain may bind to DNA as monomers or as homo- and/or heterodimers, in a sequence-specific manner. The POU family members are transcriptional regulators, many of which are known to control cell type-specific differentiation pathways. This gene is a tumor suppressor involved in Wilms tumor (WT) predisposition. Alternatively spliced transcript variants encoding distinctMore Product Information