Tisk…

Anti-BCL2L2 Rabbit Polyclonal Antibody (Cy7®)

Dodavatel: Bioss
BS-5903R-CY7
BOSSBS-5903R-CY7EA 25630 CZK
BOSSBS-5903R-CY7
Anti-BCL2L2 Rabbit Polyclonal Antibody (Cy7®)
Protilátky
Promotes cell survival. Blocks dexamethasone-induced apoptosis. Mediates survival of postmitotic Sertoli cells by suppressing death-promoting activity of BAX.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BCL2L2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Produktová specifikace

Symbol antigenu BCL2L2
Název antigenu BCL2-Like 2
Environmentally Preferable
Synonyma antigenu PPP1R51|BCL2-L-2|BCL-W|BCLW
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Cy7®
Reaktivita Human,Rat,Mouse
Hostitel Rabbit
Identifikační číslo genu 599
Izotyp IgG
Imunochemie Yes
Formulář liquid
Čištění Purified by Protein A
Koncentrace 1 μg/μl
Křížová adsorbce No
Skladovací pufr Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Skladovací teplota Store at 4°C for 12 months
Transportní teplota 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR