Tisk…

Anti-ADAMTSL3 Rabbit Polyclonal Antibody (Cy5®)

Dodavatel: Bioss
BS-5863R-CY5
BOSSBS-5863R-CY5EA 25430 CZK
BOSSBS-5863R-CY5
Anti-ADAMTSL3 Rabbit Polyclonal Antibody (Cy5®)
Protilátky
Expressed in epithelial cells of the colon, fallopian tube, skin, breast, prostate, epididymis, liver, pancreatic islets and bile ducts, as well as by vascular endothelial cells, smooth muscle cells, fibroblasts, cortical and ganglionic neurons and cardiac myocytes. Also expressed by malignant epithelial cells in colon cancer, as well as breast, prostate, renal and skin tumors. Expression is significantly reduced in colon cancer compared to normal colon.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ADAMTSL3
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host:More Product Information
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR