Tisk…

Anti-SMAGP Rabbit Polyclonal Antibody

Dodavatel: Bioss
BS-5844R
BOSSBS-5844REA 20650 CZK
BOSSBS-5844R
Anti-SMAGP Rabbit Polyclonal Antibody
Protilátky
May play a role in epithelial cell-cell contacts. May play a role in tumor invasiveness and metastasis formation.

WB(1:100-1000), FCM(1:20-100), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: SMAGP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR