Tisk…

Anti-CRNN Rabbit Polyclonal Antibody (Cy5.5®)

Dodavatel: Bioss
BS-5833R-CY5.5
BOSSBS-5833R-CY5.5EA 25630 CZK
BOSSBS-5833R-CY5.5
Anti-CRNN Rabbit Polyclonal Antibody (Cy5.5®)
Protilátky
This gene encodes a member of the "fused gene" family of proteins, which contain N terminus EF hand domains and multiple tandem peptide repeats. The encoded protein contains two EF hand Ca2+ binding domains in its N terminus and two glutamine and threonine rich 60 amino acid repeats in its C terminus. This gene, also known as squamous epithelial heat shock protein 53, may play a role in the mucosal/epithelial immune response and epidermal differentiation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CRNN
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR