Tisk…
Anti-RPS6KB1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Dodavatel: Bioss
BS-5673R-A647
BOSSBS-5673R-A647EA 26260 CZK
BOSSBS-5673R-A647
Anti-RPS6KB1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Protilátky
This gene encodes a member of the ribosomal S6 kinase family of serine/threonine kinases. The encoded protein responds to mTOR (mammalian target of rapamycin) signaling to promote protein synthesis, cell growth, and cell proliferation. Activity of this gene has been associated with human cancer. Alternatively spliced transcript variants have been observed. The use of alternative translation start sites results in isoforms with longer or shorter N-termini which may differ in their subcellular localizations. There are two pseudogenes for this gene on chromosome 17. [provided by RefSeq, Jan 2013More Product Information