Tisk…
Anti-RPS6KB1 Rabbit Polyclonal Antibody (Cy3®)
Dodavatel: Bioss
BS-5672R-CY3
BOSSBS-5672R-CY3EA 38440 CZK
BOSSBS-5672R-CY3
Anti-RPS6KB1 Rabbit Polyclonal Antibody (Cy3®)
Protilátky
This gene encodes a member of the ribosomal S6 kinase family of serine/threonine kinases. The encoded protein responds to mTOR (mammalian target of rapamycin) signaling to promote protein synthesis, cell growth, and cell proliferation. Activity of this gene has been associated with human cancer. Alternatively spliced transcript variants have been observed. The use of alternative translation start sites results in isoforms with longer or shorter N-termini which may differ in their subcellular localizations. There are two pseudogenes for this gene on chromosome 17. [provided by RefSeq, Jan 2013More Product Information
Objednat
Produktová specifikace
Symbol antigenu RPS6KB1
Název antigenu Ribosomal Protein S6 Kinase, 70kDa, Polypeptide 1
Environmentally Preferable
Synonyma antigenu PS6K|S6K-beta-1|S6K|p70(S6K)-alpha|S6K1|STK14A|p70 S6KA|p70-alpha|p70-S6K
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Cy3®
Reaktivita Human,Rat,Mouse
Hostitel Rabbit
Identifikační číslo genu 6198
Izotyp IgG
Imunochemie Yes
Modifikace phospho-Ser421
Formulář liquid
Čištění Purified by Protein A
Koncentrace 1 μg/μl
Křížová adsorbce No
Skladovací pufr Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Skladovací teplota Store at 4°C for 12 months
Transportní teplota 4°C