Tisk…
Anti-PRKDC Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Dodavatel: Bioss
BS-5130R-A647
BOSSBS-5130R-A647EA 25430 CZK
BOSSBS-5130R-A647
Anti-PRKDC Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Protilátky
Serine/threonine-protein kinase that acts as a molecular sensor for DNA damage. Involved in DNA non-homologous end joining (NHEJ) required for double-strand break (DSB) repair and V(D)J recombination. Must be bound to DNA to express its catalytic properties. Promotes processing of hairpin DNA structures in V(D)J recombination by activation of the hairpin endonuclease artemis (DCLRE1C). The assembly of the DNA-PK complex at DNA ends is also required for the NHEJ ligation step. Required to protect and align broken ends of DNA. May also act as a scaffold protein to aid the localization of DNAMore Product Information
Objednat
Produktová specifikace
Symbol antigenu PRKDC
Název antigenu Protein Kinase, DNA-Activated, Catalytic Polypeptide
Environmentally Preferable
Synonyma antigenu DNA-PKcs|IMD26|DNPK1|XRCC7|DNAPK|HYRC1|HYRC|p350
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Alexa Fluor® 647
Reaktivita Human,Rat,Mouse
Hostitel Rabbit
Identifikační číslo genu 5591
Izotyp IgG
Imunochemie Yes
Imunogen 460-510/832
Formulář liquid
Čištění Purified by Protein A
Koncentrace 1 μg/μl
Křížová adsorbce No
Skladovací pufr Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Skladovací teplota Store at 4°C for 12 months
Transportní teplota 4°C