Tisk…

Anti-IDI1 Rabbit Polyclonal Antibody (Cy5®)

Dodavatel: Bioss
BS-5073R-CY5
BOSSBS-5073R-CY5EA 25630 CZK
BOSSBS-5073R-CY5
Anti-IDI1 Rabbit Polyclonal Antibody (Cy5®)
Protilátky
IDI1 is a peroxisomally localized enzyme that catalyzes the interconversion of isopentenyl diphosphate (IPP) to its highly electrophilic isomer, dimethylallyl diphosphate (DMAPP). These are substrates for the reactions that ultimately result in the synthesis of cholesterol. There is reduction in IPP isomerase activity in peroxisomal deficiency diseases such as Zellweger syndrome and neonatal adrenoleukodystrophy.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IDI1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Produktová specifikace

Symbol antigenu IDI1
Environmentally Preferable
Název antigenu Isopentenyl-Diphosphate Delta Isomerase 1
Synonyma antigenu IPPI1|IPP1
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Cy5®
Reaktivita Human,Rat,Mouse
Hostitel Rabbit
Identifikační číslo genu 3422
Izotyp IgG
Imunochemie Yes
Formulář liquid
Čištění Purified by Protein A
Koncentrace 1 μg/μl
Křížová adsorbce No
Skladovací pufr Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Skladovací teplota Store at 4°C for 12 months
Transportní teplota 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR