Tisk…

Anti-NMRK1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Dodavatel: Bioss
BS-5058R-A647
BOSSBS-5058R-A647EA 25430 CZK
BOSSBS-5058R-A647
Anti-NMRK1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Protilátky
ECHS1 (Enoyl Coenzyme A hydratase, short chain, 1) catalyzes the hydration of 2-trans-enoyl-coenzyme A (CoA) intermediates to L-3-hydroxyacyl-CoAs, in the second step of the mitochondrial fatty acid beta-oxidation pathway.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NMRK1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Produktová specifikace

Symbol antigenu NMRK1
Název antigenu Nicotinamide Riboside Kinase 1
Environmentally Preferable
Synonyma antigenu NRK1|bA235O14.2|C9orf95
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Alexa Fluor® 647
Reaktivita Human,Rat,Mouse
Hostitel Rabbit
Identifikační číslo genu 54981
Izotyp IgG
Imunochemie Yes
Formulář liquid
Čištění Purified by Protein A
Koncentrace 1 μg/μl
Křížová adsorbce No
Skladovací pufr Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Skladovací teplota Store at 4°C for 12 months
Transportní teplota 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR