Tisk…

Anti-CYP4A11 Rabbit Polyclonal Antibody (Cy7®)

Dodavatel: Bioss
BS-5054R-CY7
BOSSBS-5054R-CY7EA 38440 CZK
BOSSBS-5054R-CY7
Anti-CYP4A11 Rabbit Polyclonal Antibody (Cy7®)
Protilátky
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and hydroxylates medium-chain fatty acids such as laurate and myristate. [provided by RefSeq, Jul 2008].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CYP4A11
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR