Tisk…
Anti-BDH1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Dodavatel: Bioss
BS-5041R-FITC
BOSSBS-5041R-FITCEA 25630 CZK
BOSSBS-5041R-FITC
Anti-BDH1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Protilátky
This gene encodes a member of the short-chain dehydrogenase/reductase gene family. The encoded protein forms a homotetrameric lipid-requiring enzyme of the mitochondrial membrane and has a specific requirement for phosphatidylcholine for optimal enzymatic activity. The encoded protein catalyzes the interconversion of acetoacetate and (R)-3-hydroxybutyrate, the two major ketone bodies produced during fatty acid catabolism. Alternatively spliced transcript variants encoding the same protein have been described. [provided by RefSeq, Jul 2008].
Recommended Dilutions: IF(IHC-P): 1:50-200