Tisk…

Anti-PYGL Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Dodavatel: Bioss
BS-5011R-A750
BOSSBS-5011R-A750EA 25340 CZK
BOSSBS-5011R-A750
Anti-PYGL Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Protilátky
Phosphorylase is an important allosteric enzyme in carbohydrate metabolism. Enzymes from different sources differ in their regulatory mechanisms and in their natural substrates. However, all known phosphorylases share catalytic and structural properties.

Type: Primary
Antigen: PYGL
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 451-550/847
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Produktová specifikace

Environmentally Preferable
Název antigenu PYGL
Synonyma antigenu GSD6 antibody Hers disease, glycogen storage disease type VI|Phosphorylase glycogen liver|Glycogen phosphorylase liver form|PYGL_HUMAN|Glycogen phosphorylase liver
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Alexa Fluor® 750
Reaktivita
Hostitel Rabbit
Identifikační číslo genu 5836
Izotyp IgG
Imunochemie Yes
ImunoFluorescence Yes
Modifikace Unmodified
Epitop 451-550/847
Imunogen KLH conjugated synthetic peptide derived from human PYGL
Formulář Liquid
Molekulární hmotnost 97 kDa
Čištění Purified by Protein A
Koncentrace 1 µg/µl
Skladovací pufr Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Skladovací teplota Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Transportní teplota 4 °C
Testované aplikace ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR