Tisk…

Anti-LEP Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Dodavatel: Bioss
BS-4604R-A350
BOSSBS-4604R-A350EA 25430 CZK
BOSSBS-4604R-A350
Anti-LEP Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Protilátky
Leptin, the product of the ob (obese) gene, is a 16kD protein consisting of 146 amino acid residues. Leptin is produced in the adipose tissue, and is considered to play an important role in appetite control, fat metabolism and regulation of body weight. It targets the central nervous system, particularly hypothalamus, affecting food intake. Leptin levels are high in most obese individuals. Studies have shown that it may also influence reproductive function.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LEP
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
More Product Information
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR