Tisk…
Anti-MMP2 Rabbit Polyclonal Antibody (Cy7®)
Dodavatel: Bioss
BS-4599R-CY7
BOSSBS-4599R-CY7EA 25890 CZK
BOSSBS-4599R-CY7
Anti-MMP2 Rabbit Polyclonal Antibody (Cy7®)
Protilátky
Ubiquitinous metalloproteinase that is involved in diverse functions such as remodeling of the vasculature, angiogenesis, tissue repair, tumor invasion, inflammation, and atherosclerotic plaque rupture. As well as degrading extracellular matrix proteins, can also act on several nonmatrix proteins such as big endothelial 1 and beta-type CGRP promoting vasoconstriction. Also cleaves KISS at a Gly-|-Leu bond. Appears to have a role in myocardial cell death pathways. Contributes to myocardial oxidative stress by regulating the activity of GSK3beta. Cleaves GSK3beta in vitro. Involved in theMore Product Information