Tisk…

Anti-ASGR1 Rabbit Polyclonal Antibody

Dodavatel: Bioss
BS-4119R
BOSSBS-4119REA 20650 CZK
BOSSBS-4119R
Anti-ASGR1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ASGR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR