Tisk…

Anti-GK Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Dodavatel: Bioss
BS-4062R-A350
BOSSBS-4062R-A350EA 25430 CZK
BOSSBS-4062R-A350
Anti-GK Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Protilátky
Glycerol kinase catalyzes the formation of glycerol 3 phosphate from ATP and glycerol. Dihydroxyacetone and L glyceraldehyde can also act as acceptors; UTP and, in the case of the yeast enzyme, ITP and GTP can act as donors. It provides a way for glycerol derived from fats or glycerides to enter the glycolytic pathway.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Glycerol kinase
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR