Tisk…

Anti-ZBTB8A Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Dodavatel: Bioss
BS-4057R-A555
BOSSBS-4057R-A555EA 25430 CZK
BOSSBS-4057R-A555
Anti-ZBTB8A Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Protilátky
DEGS1 is a member of the membrane fatty acid desaturase family which is responsible for inserting double bonds into specific positions in fatty acids. It contains three His containing consensus motifs that are characteristic of a group of membrane fatty acid desaturases. It has sphingolipid-delta-4-desaturase activity and converts D-erythro-sphinganine to D-erythro-sphingosine (E-sphing-4-enine).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZBTB8A
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Produktová specifikace

Symbol antigenu ZBTB8A
Environmentally Preferable
Název antigenu Zinc Finger And BTB Domain Containing 8A
Synonyma antigenu ZBTB8|ZNF916A|BOZF1
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Alexa Fluor® 555
Reaktivita Human,Rat,Mouse
Hostitel Rabbit
Identifikační číslo genu 653121
Izotyp IgG
Imunochemie Yes
Formulář liquid
Čištění Purified by Protein A
Koncentrace 1 μg/μl
Křížová adsorbce No
Skladovací pufr Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Skladovací teplota Store at 4°C for 12 months
Transportní teplota 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR