Tisk…

Anti-ACADM Rabbit Polyclonal Antibody (Cy5.5®)

Dodavatel: Bioss
BS-4047R-CY5.5
BOSSBS-4047R-CY5.5EA 25630 CZK
BOSSBS-4047R-CY5.5
Anti-ACADM Rabbit Polyclonal Antibody (Cy5.5®)
Protilátky
ACADM protein is a medium chain specific (C4 to C12 straight chain) acyl Coenzyme A dehydrogenase. The enzyme catalyzes the initial step of the mitochondrial fatty acid beta oxidation pathway. ACADM expression is induced during periods of fasting, when reliance on fatty acids for energy is increased. Clinical phenotypes are associated with ACADM hereditary deficiency.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ACADM
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR