Tisk…
Anti-PHKB Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Dodavatel: Bioss
BS-4032R-A680
BOSSBS-4032R-A680EA 24890 CZK
BOSSBS-4032R-A680
Anti-PHKB Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Protilátky
Phosphorylase kinase is a polymer of 16 subunits, four each of alpha, beta, gamma and delta. The alpha subunit includes the skeletal muscle and hepatic isoforms, encoded by two different genes. The beta subunit is the same in both the muscle and hepatic isoforms, encoded by this gene, which is a member of the phosphorylase b kinase regulatory subunit family. The gamma subunit also includes the skeletal muscle and hepatic isoforms, encoded by two different genes. The delta subunit is a calmodulin and can be encoded by three different genes. The gamma subunits contain the active site of theMore Product Information