Tisk…

Anti-SDHD Rabbit Polyclonal Antibody (Cy7®)

Dodavatel: Bioss
BS-3971R-CY7
BOSSBS-3971R-CY7EA 25630 CZK
BOSSBS-3971R-CY7
Anti-SDHD Rabbit Polyclonal Antibody (Cy7®)
Protilátky
Membrane-anchoring subunit of succinate dehydrogenase (SDH) that is involved in complex II of the mitochondrial electron transport chain and is responsible for transferring electrons from succinate to ubiquinone (coenzyme Q).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SDHD
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR