Tisk…
Anti-CCS Rabbit Polyclonal Antibody (Cy7®)
Dodavatel: Bioss
BS-3896R-CY7
BOSSBS-3896R-CY7EA 25630 CZK
BOSSBS-3896R-CY7
Anti-CCS Rabbit Polyclonal Antibody (Cy7®)
Protilátky
Superoxide dismutase (SOD) is an antioxidant enzyme involved in the defense system against reactive oxygen species (ROS). SOD catalyzes the dismutation reaction of superoxide radical anion (O2-) to hydrogen peroxide, which is then catalyzed to innocuous O2 and H2O by glutathione peroxidase and catalase. Several classes of SOD have been identified. These include intracellular copper, zinc SOD (Cu, Zn-SOD/SOD-1), mitochondrial manganese SOD (Mn-SOD/SOD-2) and extracellular Cu, Zn-SOD (EC-SOD/SOD-3). SOD1 is found in all eukaryotic species as a homodimeric 32 kDa enzyme containing one each ofMore Product Information