Tisk…

Anti-GPX1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Dodavatel: Bioss
BS-3882R-A680
BOSSBS-3882R-A680EA 25600 CZK
BOSSBS-3882R-A680
Anti-GPX1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Protilátky
This gene product belongs to the family of glutathione peroxidase, which functions in the detoxification of hydrogen peroxide. It contains a selenocysteine (Sec) residue at its active site. The selenocysteine is encoded by the UGA codon, which normally signals translation termination. The 3' UTR of Sec-containing genes have a common stem-loop structure, the sec insertion sequence (SECIS), which is necessary for the recognition of UGA as a Sec codon rather than as a stop signal.

Type: Primary
Antigen: GPX1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 51-150More Product Information
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR