Tisk…

Anti-ATG4A Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Dodavatel: Bioss
BS-3873R-A680
BOSSBS-3873R-A680EA 25340 CZK
BOSSBS-3873R-A680
Anti-ATG4A Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Protilátky
ATG4A is a cysteine protease required for autophagy, which cleaves the C-terminal part of either MAP1LC3, GABARAPL2 or GABARAP, allowing the liberation of form I. A subpopulation of form I is subsequently converted to a smaller form (form II). Form II, with a revealed C-terminal glycine, is considered to be the phosphatidylethanolamine (PE)-conjugated form, and has the capacity for the binding to autophagosomes.

Type: Primary
Antigen: ATG4A
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 51-150/398
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR