Tisk…

Anti-STS Rabbit Polyclonal Antibody

Dodavatel: Bioss
BS-3857R
BOSSBS-3857REA 20650 CZK
BOSSBS-3857R
Anti-STS Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: STS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR