Tisk…

Anti-SCD Rabbit Polyclonal Antibody (Cy5.5®)

Dodavatel: Bioss
BS-3787R-CY5.5
BOSSBS-3787R-CY5.5EA 25630 CZK
BOSSBS-3787R-CY5.5
Anti-SCD Rabbit Polyclonal Antibody (Cy5.5®)
Protilátky
Terminal component of the liver microsomal stearyl-CoA desaturase system, that utilizes O(2) and electrons from reduced cytochrome b5 to catalyze the insertion of a double bond into a spectrum of fatty acyl-CoA substrates including palmitoyl-CoA and stearoyl-CoA.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SCD
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Produktová specifikace

Symbol antigenu SCD
Název antigenu Stearoyl-CoA Desaturase
Environmentally Preferable
Synonyma antigenu PRO1933|stearoyl-CoA desaturase opposite strand|MSTP008|FADS5|SCDOS|delta(9)-desaturase|predicted protein of HQ0998|delta-9-desaturase|delta-9 desaturase|fatty acid desaturase|SCD1|acyl-CoA desaturase
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Cy5.5®
Reaktivita Human,Rat,Mouse
Hostitel Rabbit
Identifikační číslo genu 6319
Izotyp IgG
Imunochemie Yes
Imunogen 310-359/359
Formulář liquid
Čištění Purified by Protein A
Koncentrace 1 μg/μl
Křížová adsorbce No
Skladovací pufr Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Skladovací teplota Store at 4°C for 12 months
Transportní teplota 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR