Tisk…
Anti-PP1A/PP2CA/PPP1CA Rabbit Polyclonal Antibody (Cy3®)
Dodavatel: Bioss
BS-3756R-CY3
BOSSBS-3756R-CY3EA 25630 CZK
BOSSBS-3756R-CY3
Anti-PP1A/PP2CA/PPP1CA Rabbit Polyclonal Antibody (Cy3®)
Protilátky
PPP1A is a serine/threonine protein phosphatase catalytic subunit that is essential for regulating cellular stress responses in eukaryotes. It binds to magnesium or manganese ions and exists as a monomer. It is essential for cell division, and participates in the regulation of glycogen metabolism, muscle contractility and protein synthesis. PPP1A is involved in the regulation long term synaptic plasticity and may play an important role in dephosphorylating substrates such as Ca2+/calmodulin dependent protein kinase II.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PP1More Product Information
Objednat
Produktová specifikace
Symbol antigenu PP1A/PP2CA/PPP1CA
Environmentally Preferable
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Cy3®
Reaktivita Human,Rat,Mouse
Hostitel Rabbit
Identifikační číslo genu 5499
Izotyp IgG
Imunochemie Yes
Formulář liquid
Čištění Purified by Protein A
Koncentrace 1 μg/μl
Křížová adsorbce No
Skladovací pufr Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Skladovací teplota Store at 4°C for 12 months
Transportní teplota 4°C