Tisk…
Anti-S6K1 Rabbit Polyclonal Antibody
Dodavatel: Bioss
BS-3700R
BOSSBS-3700REA 21160 CZK
BOSSBS-3700R
Anti-S6K1 Rabbit Polyclonal Antibody
Protilátky
This gene encodes a member of the ribosomal S6 kinase family of serine/threonine kinases. The encoded protein responds to mTOR (mammalian target of rapamycin) signaling to promote protein synthesis, cell growth, and cell proliferation. Activity of this gene has been associated with human cancer. Alternatively spliced transcript variants have been observed. The use of alternative translation start sites results in isoforms with longer or shorter N-termini which may differ in their subcellular localizations. There are two pseudogenes for this gene on chromosome 17. [provided by RefSeq, Jan 2013More Product Information