Tisk…

Anti-DIO2 Rabbit Polyclonal Antibody

Dodavatel: Bioss
BS-3673R
BOSSBS-3673REA 20650 CZK
BOSSBS-3673R
Anti-DIO2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: DIO2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR