Tisk…
Anti-CAM Rabbit Polyclonal Antibody (Cy3®)
Dodavatel: Bioss
BS-3666R-CY3
BOSSBS-3666R-CY3EA 25630 CZK
BOSSBS-3666R-CY3
Anti-CAM Rabbit Polyclonal Antibody (Cy3®)
Protilátky
Calmodulin consists of two glycoproteins, 34 and 39 kDa, sometimes designated epithelial antigen, epithelial specific antigen, and epithelial glycoprotein. The glycoproteins are located on the cell membrane surface and in the cytoplasm of virtually all epithelial cells with the exception of most squamous epithelia, hepatocytes, renal proximal tubular cells, gastric parietal cells and myoepithelial cells. Epithelial Calmodulin is found in the large majority of adenocarcinomas of most sites (50-100% in various studies; as well as neuroendocrine tumours, including small cell carcinoma. RenalMore Product Information