Tisk…

Anti-NCF4 Rabbit Polyclonal Antibody

Dodavatel: Bioss
BS-3497R
BOSSBS-3497REA 21160 CZK
BOSSBS-3497R
Anti-NCF4 Rabbit Polyclonal Antibody
Protilátky
NCF4 is a cytosolic oxidase associated protein. It is a component of the nicotinamide adenine dinucleotide phosphate oxidase, which mediates down regulation of NADPH oxidase through interactions with its SH3 domain. NCF4 is associated with p67 phox but is absent in patients with chronic granulomatous disease who lack p67 phox.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: NCF4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR