Tisk…
Anti-CDC42 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Dodavatel: Bioss
BS-3369R-A488
BOSSBS-3369R-A488EA 26230 CZK
BOSSBS-3369R-A488
Anti-CDC42 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Protilátky
The protein encoded by this gene is a small GTPase of the Rho-subfamily, which regulates signaling pathways that control diverse cellular functions including cell morphology, migration, endocytosis and cell cycle progression. This protein is highly similar to Saccharomyces cerevisiae Cdc 42, and is able to complement the yeast cdc42-1 mutant. The product of oncogene Dbl was reported to specifically catalyze the dissociation of GDP from this protein. This protein could regulate actin polymerization through its direct binding to Neural Wiskott-Aldrich syndrome protein (N-WASP), whichMore Product Information