Tisk…
Anti-PLCG2 Rabbit Polyclonal Antibody (Cy5.5®)
Dodavatel: Bioss
BS-3338R-CY5.5
BOSSBS-3338R-CY5.5EA 26460 CZK
BOSSBS-3338R-CY5.5
Anti-PLCG2 Rabbit Polyclonal Antibody (Cy5.5®)
Protilátky
Enzymes of the phospholipase C family catalyze the hydrolysis of phospholipids to yield diacylglycerols and water soluble phosphorylated derivatives of the lipid head groups. A number of these enzymes have specificity for phosphoinositides. Of the phosphoinositide specific phospholipase C enzymes, C beta is regulated by heterotrimeric G protein coupled receptors, while the closely related C gamma 1 and C gamma 2 enzymes are controlled by receptor tyrosine kinases. The C gamma 1 and C gamma 2 enzymes are composed of phospholipase domains that flank regions of homology to noncatalytic domainsMore Product Information