Tisk…
Anti-GRIN1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Dodavatel: Bioss
BS-3302R-A350
BOSSBS-3302R-A350EA 26230 CZK
BOSSBS-3302R-A350
Anti-GRIN1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Protilátky
Neuronal Marker The protein encoded by this gene is a critical subunit of N-methyl-D-aspartate receptors, members of the glutamate receptor channel superfamily which are heteromeric protein complexes with multiple subunits arranged to form a ligand-gated ion channel. These subunits play a key role in the plasticity of synapses, which is believed to underlie memory and learning. Cell-specific factors are thought to control expression of different isoforms, possibly contributing to the functional diversity of the subunits. Alternatively spliced transcript variants have been described. [More Product Information