Tisk…

Anti-Factor 8 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Dodavatel: Bioss
BS-2974R-A680
BOSSBS-2974R-A680EA 25600 CZK
BOSSBS-2974R-A680
Anti-Factor 8 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Protilátky
Factor VIII, along with calcium and phospholipid, acts as a cofactor for factor IXa when it converts factor X to the activated form, factor Xa.

Type: Primary
Antigen: Factor 8
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 85-190/2351
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR